1,3,5-Triethylbenzene ≥97%

Warning

132071-5G 132071-25G
220036-071EA 76.57 USD
220036-071 220036-070
1,3,5-Triethylbenzene ≥97%
1,3,5-Triethylbenzene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.8631 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009261
CAS Number: 102-25-0
EINECS: 203-017-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR