p-Toluenesulfonyl hydrazide ≥97%

Synonyms: 4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide

132004-25G 132004-500G 132004-100G
220036-058EA 56.86 USD
220036-058 220036-059 220036-057
p-Toluenesulfonyl hydrazide ≥97%
p-Toluenesulphonyl hydrazide
Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 105…108 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007588
CAS Number: 1576-35-8
EINECS: 216-407-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR