Tetraethyl orthosilicate (TEOS) ≥98%, Reagent Grade

Danger

Synonyms: Tetraethoxysilane

131903-4L 131903-1L 131903-25ML 131903-500ML
220035-991EA 346.15 USD
220035-991 220035-989 220035-990 220035-992
Tetraethyl orthosilicate (TEOS) ≥98%, Reagent Grade
Tetraethyl orthosilicate (TEOS)
Formula: Si(OC₂H₅)₄
MW: 208.33 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: -85.5 °C
Density: 0.933 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR