Sebacoyl dichloride ≥92%, Technical Grade

Danger

Synonyms: Sebacoyl chloride

131784-500G 131784-250G 131784-50G
220035-974EA 394.92 USD
220035-974 220035-973 220035-975
Sebacoyl dichloride ≥92%, Technical Grade
Sebacoyl dichloride
Formula: C₁₀H₁₆Cl₂O₂
MW: 239.14 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: -5 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR