Sarcosine ≥98%

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

131776-500G 131776-100G
220035-972EA 219.08 USD
220035-972 220035-731
Sarcosine ≥98%
Sarcosine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR