4-Phenyl-3-thiosemicarbazide ≥99%

Synonyms: 4-Phenyl-3-thiosemicarbazide, N-Phenylhydrazinecarbothioamide, 4-Phenylthiosemicarbazide

131482-50G 131482-10G
220033-224EA 224 USD
220033-224 220033-715
4-Phenyl-3-thiosemicarbazide ≥99%
4-Phenyl-3-thiosemicarbazide
Formula: C₇H₉N₃S
MW: 167.23 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00007615
CAS Number: 5351-69-9
EINECS: 226-329-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR