1-Nonanol ≥98%

Warning

Synonyms: Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol

131210-500ML 131210-100ML
220035-857EA 105.43 USD
220035-857 220035-856
1-Nonanol ≥98%
1-Nonanol
Formula: H₃C(CH₂)₇CH₂OH
MW: 144.26 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: -7 °C
Density: 0.8419 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00002990
CAS Number: 143-08-8
EINECS: 205-583-7
UN: 3082
ADR: 9,III
Merck Index: 12,06774

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR