Indigo carmine, certified by the Biological Stain Commission

Warning

Synonyms: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

131164-100G 131164-25G
220035-849EA 252 USD
220035-849 220035-850
Indigo carmine, certified by the Biological Stain Commission
Indigo carmine
Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR