3-Butyne-1-ol ≥97%

Danger

Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

130850-25G 130850-5G 130850-100G
220035-804EA 145.38 USD
220035-804 220035-805 220035-803
3-Butyne-1-ol ≥97%
3-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 128.8 °C (1013 hPa)
Melting Pt: -64 °C
Density: 0.9262 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR