Isobutyrophenone ≥97%

Synonyms: Isopropyl phenyl ketone, 2-Methyl-1-phenyl-1-propanone, Phenyl isopropyl ketone, α-Methylpropiophenone

130362-100G 130362-25G
220035-782EA 97.25 USD
220035-782 220035-783
Isobutyrophenone ≥97%
Isobutyrophenone
Formula: (C₆H₅)C(O)CH(CH₃)₂
MW: 148.2 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 1 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008917
CAS Number: 611-70-1
EINECS: 210-275-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR