(2-Nitrophenyl)acetic acid ≥98%

Synonyms: 2-(2-Nitrophenyl)acetic acid

130249-100G 130249-25G
220035-704EA 176.62 USD
220035-704 220035-705
(2-Nitrophenyl)acetic acid ≥98%
(2-Nitrophenyl)acetic acid
Formula: (C₆H₄NO₂)CH₂COOH
MW: 181.15 g/mol
Melting Pt: 136…139 °C
MDL Number: MFCD00007190
CAS Number: 3740-52-1
EINECS: 223-128-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR