5-Nitroindoline

Warning

Synonyms: 2,3-Dihydro-5-nitro-(1H)-indole

130214-5G 130214-25G
220035-632EA 564.55 USD
220035-632 220035-631
5-Nitroindoline
5-Nitroindoline
Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
MDL Number: MFCD00005709
CAS Number: 32692-19-6
EINECS: 251-158-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR