4-Nitrobenzaldehyde ≥98%

130176-10G 130176-100G 130176-25G
220034-958EA 40.71 USD
220034-958 220034-957 220034-959
4-Nitrobenzaldehyde ≥98%
4-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR