2-Naphthol

Warning

Synonyms: 2-Hydroxynaphthalene, β-Naphthol, β-Hydroxynaphthalene

130109-1KG 130109-250G
220035-624EA 75.28 USD
220035-624 220034-955
2-Naphthol
2-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 121.6 °C
Density: 1.217 g/cm³ (20 °C)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III
Merck Index: 12,06471

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR