(Methyl)triphenylphosphonium bromide

130079-25G 130079-100G 130079-500G
220035-620EA 34.14 USD
220035-620 220035-619 220035-621
(Methyl)triphenylphosphonium bromide
(Methyl)triphenylphosphonium bromide
Formula: C₁₉H₁₈BrP
MW: 357.23 g/mol
Melting Pt: 232 °C
Flash Pt: >240 °C
MDL Number: MFCD00011804
CAS Number: 1779-49-3
EINECS: 217-218-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR