4-Methyl-3-thiosemicarbazide ≥97%

130060-10G 130060-50G
220035-015EA 74.74 USD
220035-015 220035-016
4-Methyl-3-thiosemicarbazide ≥97%
4-Methyl-3-thiosemicarbazide
Formula: C₂H₇N₃S
MW: 105.16 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 136 °C
Density: 1.362 g/cm³ (20 °C)
MDL Number: MFCD00007617
CAS Number: 6610-29-3
EINECS: 229-563-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR