1-Methylpiperazine

130001-500G 130001-100G 130001-5G
220035-618EA 157.23 USD
220035-618 220035-617 220035-007
1-Methylpiperazine
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.9085 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR