Methyl methanesulfonate ≥99%

129925-25G 129925-100G 129925-5G
220035-611EA 149.69 USD
220035-611 220035-610 220035-612
Methyl methanesulfonate ≥99%
Methyl methanesulphonate
Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00007557
CAS Number: 66-27-3
EINECS: 200-625-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,06170

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR