2-Methyl-3-butyne-2-ol ≥98%

Synonyms: 3-Hydroxy-3-methyl-1-butyne, 2-Methylbut-3-yn-2-ol

129763-1L 129763-5ML 129763-100ML
220035-586EA 99.83 USD
220035-586 220035-587 220035-585
2-Methyl-3-butyne-2-ol ≥98%
2-Methyl-3-butyne-2-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 104 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004467
CAS Number: 115-19-5
EINECS: 204-070-5
UN: 1993
ADR: 3,II
Merck Index: 12,06113

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR