Indole-3-carbaldehyd

Synonyms: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

129445-25G 129445-5G
220035-491EA 89.92 USD
220035-491 220035-492
Indole-3-carbaldehyd
Indole-3-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 192…195 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR