4-Iodoaniline ≥98%

Warning

129364-25G 129364-100G
220035-212EA 61.06 USD
220035-212 220035-211
4-Iodoaniline ≥98%
4-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 61 - 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007848
CAS Number: 540-37-4
EINECS: 208-743-4
Merck Index: 13,05049

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR