2,3-Dimethyl-2-butene ≥98%

Synonyms: Tetramethylethylene

129259-25ML 129259-100ML 129259-1L
220033-675EA 101.23 USD
220033-675 220033-673 220033-674
2,3-Dimethyl-2-butene ≥98%
2,3-Dimethyl-2-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 73 °C (1013 hPa)
Melting Pt: –75 °C
Density: 0.7188 g/cm³ (20 °C)
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR