2,3-Dimethyl-2-butanol ≥98%

129240-25G 129240-5G
220033-671EA 614.92 USD
220033-671 220033-672
2,3-Dimethyl-2-butanol ≥98%
2,3-Dimethyl-2-butanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 119 °C (1013 hPa)
Melting Pt: –10.4 °C
Density: 0.8387 g/cm³ (0 °C)
MDL Number: MFCD00004463
CAS Number: 594-60-5
EINECS: 209-847-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR