N,O-Bis(trimethylsilyl)acetamide for synthesis

Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

128910-25ML 128910-100ML 128910-10ML 128910-1L
220035-192EA 50.94 USD
220035-192 220033-684 220033-685 220033-686
N,O-Bis(trimethylsilyl)acetamide for synthesis
N,O-Bis(trimethylsilyl)acetamide
Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: ∼24 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR