3,5-Dinitrosalicylic acid ≥98%

Warning

Synonyms: 2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid

128848-5G 128848-500G 128848-100G
220033-683EA 19.82 USD
220033-683 220033-682 220033-681
3,5-Dinitrosalicylic acid ≥98%
3,5-Dinitrosalicylic acid
Formula: C₇H₄N₂O₇
MW: 228.12 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00007104
CAS Number: 609-99-4
EINECS: 210-204-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR