Piperidin-4-ol

Warning

Synonyms: 4-Piperidinol, 4-Hydroxypiperidine

128775-5G 128775-100G 128775-25G
220035-489EA 37.69 USD
220035-489 220035-487 220035-488
Piperidin-4-ol
Piperidin-4-ol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 86…89 °C
Flash Pt: <100 °C
MDL Number: MFCD00005999
CAS Number: 5382-16-1
EINECS: 226-373-1
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR