6-Methyl-2-pyridone

Warning

Synonyms: 2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one

128740-5G 128740-25G
220035-484EA 51.69 USD
220035-484 220034-813
6-Methyl-2-pyridone
6-Methyl-2-pyridone
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 159…160 °C
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR