trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

128708-100G 128708-5G 128708-25G
220034-810EA 233.69 USD
220034-810 220034-812 220034-811
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD00004400
CAS Number: 537-98-4
EINECS: 208-679-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR