2-(2-Hydroxyethyl)pyridine

Synonyms: 2-Pyridineethanol, 2-(2-Pyridyl)ethanol

128643-500G 128643-5G 128643-100G
220034-808EA 124.92 USD
220034-808 220034-809 220034-807
2-(2-Hydroxyethyl)pyridine
2-(2-Hydroxyethyl)pyridine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: -7.8 °C
Density: 1.1111 g/cm³ (4 °C)
MDL Number: MFCD00006364
CAS Number: 103-74-2
EINECS: 203-140-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR