2-Hydroxyethyl methacrylate ≥97%

Warning

Synonyms: HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate

128635-1KG 128635-500G 128635-18KG
220034-805EA 64.72 USD
220034-805 220034-806 220034-804
2-Hydroxyethyl methacrylate ≥97%
2-Hydroxyethyl methacrylate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR