(±)-2-Hexanol ≥99%

Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane

128570-25G 128570-100G
220035-482EA 95.2 USD
220035-482 220035-481
(±)-2-Hexanol ≥99%
(±)-2-Hexanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004585
CAS Number: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR