1,5-Hexadiene

Synonyms: Hexa-1,5-diene

128554-25G 128554-100G
220035-478EA 60.31 USD
220035-478 220035-477
1,5-Hexadiene
1,5-Hexadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: -141 °C
Density: 0.692 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008666
CAS Number: 592-42-7
EINECS: 209-754-7
UN: 2458
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR