Glyoxal 40% (w/w) in aqueous solution
|
Warning
|
Synonyms:
Ethandial
128465-1KG
128465-3KG
128465-100G
128465-5G
128465-22KG
220035-091EA
90.77
USD
220035-091
220035-093
220035-090
220035-094
220035-092
Glyoxal 40% (w/w) in aqueous solution
Glyoxal
Formula:
C₂H₂O₂ MW: 58.04 g/mol Density: 1.27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006957 CAS Number: 107-22-2 EINECS: 203-474-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...