4-Fluorobenzoic acid ≥98%

Warning

128384-25G 128384-5G
220035-081EA 36.18 USD
220035-081 220035-082
4-Fluorobenzoic acid ≥98%
4-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 185 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 13,04198

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR