4-Fluorobenzaldehyde ≥98%

128376-50G 128376-250G 128376-10G
220035-080EA 79.58 USD
220035-080 220035-079 220035-078
4-Fluorobenzaldehyde ≥98%
4-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: -10 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003378
CAS Number: 459-57-4
EINECS: 207-293-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR