Fluorene

Synonyms: 9H-Fluorene, Diphenylenmethane

128333-500G 128333-5G 128333-100G
220035-460EA 238 USD
220035-460 220035-461 220035-459
Fluorene
Fluorene
Formula: C₁₃H₁₀
MW: 166.22 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 113 - 115 °C
Density: 1.203 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001111
CAS Number: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9,III
Merck Index: 13,04181

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR