4-Methylbenzyl alcohol ≥98%

Synonyms: 4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol

127809-25G 127809-5G 127809-100G
220034-987EA 69.03 USD
220034-987 220034-988 220035-413
4-Methylbenzyl alcohol ≥98%
4-Methylbenzyl alcohol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 59 °C
Density: 0.978 g/cm³ (72 °C)
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR