Diethylamine hydrochloride

Synonyms: Diethylammonium chloride

127744-500G 127744-100G 127744-2.5KG
220035-409EA 82.82 USD
220035-409 220035-407 220035-408
Diethylamine hydrochloride
Diethylamine hydrochloride
Formula: C₄H₁₂ClN
MW: 109.6 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 220 - 225 °C
Density: 1.0475 g/cm³ (21 °C)
MDL Number: MFCD00012499
CAS Number: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR