2-Nitrobenzoic acid ≥95%

127698-100G 127698-5G
220035-405EA 160.46 USD
220035-405 220035-406
2-Nitrobenzoic acid ≥95%
2-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146 - 148 °C
Density: 1.575 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007137
CAS Number: 552-16-9
EINECS: 209-004-9
Merck Index: 12,06686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR