3-Aminobenzoic acid ≥98%

127671-25G 127671-100G 127671-500G
220035-403EA 33.38 USD
220035-403 220034-954 220035-404
3-Aminobenzoic acid ≥98%
3-Aminobenzoic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 174 °C
Density: 1.5105 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4
Merck Index: 13,00420

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR