2-(2-Aminoethylamino)ethanol ≥99%

Danger

Synonyms: N-(2-Hydroxyethyl)ethylenediamine, AEEA

127582-1KG 127582-100G 127582-2KG
220034-945EA 112 USD
220034-945 220034-944 220034-946
2-(2-Aminoethylamino)ethanol ≥99%
2-(2-Aminoethylamino)ethanol
Formula: C₄H₁₂N₂O
MW: 104.15 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: –28 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00008170
CAS Number: 111-41-1
EINECS: 203-867-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR