2-Methylbenzylamine ≥96%

Danger

Synonyms: o-Tolylmethanamine, 2-(Aminomethyl)toluene

127132-5G 127132-1G
220034-224EA 36.62 USD
220034-224 220034-223
2-Methylbenzylamine ≥96%
2-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: ? 0 °C
Density: 0.9921 g/cm³ (0 °C)
MDL Number: MFCD00008112
CAS Number: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR