N,N,N'-Trimethylethylenediamine ≥97%

Synonyms: N,N,N'-trimethylethane-1,2-diamine

127124-5G 127124-25G
220034-222EA 112 USD
220034-222 220034-887
N,N,N'-Trimethylethylenediamine ≥97%
N,N,N'-Trimethylethylenediamine
Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 116 °C (1013 hPa)
Density: 0.783 g/cm³ (20 °C)
MDL Number: MFCD00014874
CAS Number: 142-25-6
EINECS: 205-529-2
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR