1,2-Bis(diethylamino)ethane ≥98%

Danger

Synonyms: N1,N1,N2,N2-Tetraethylethane-1,2-diamine, N,N,N',N'-Tetraethylethylenediamine

127078-10G 127078-50G
220034-935EA 82.92 USD
220034-935 220034-936
1,2-Bis(diethylamino)ethane ≥98%
1,2-Bis(diethylamino)ethane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 189 °C (1013 hPa)
Density: 0.799 g/cm³ (25 °C)
MDL Number: MFCD00009055
CAS Number: 150-77-6
EINECS: 205-770-3
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR