1-Naphthylmethylamine

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

127035-5G 127035-25G
220034-934EA 93.37 USD
220034-934 220034-933
1-Naphthylmethylamine
1-Naphthylmethylamine
Formula: C₁₁H₁₁N
MW: 157.22 g/mol
Boiling Pt: 92 °C (0.1 mmHg)
Melting Pt: 267…269 °C
Density: 1.0958 g/cm³ (20 °C)
MDL Number: MFCD00004048
CAS Number: 118-31-0
EINECS: 204-244-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR