N-Ethylbenzylamine ≥97%

Danger

Synonyms: N-Benzylethylamine, N-Benzylethanamine

126993-100ML 126993-25ML
220035-401EA 155.08 USD
220035-401 220035-402
N-Ethylbenzylamine ≥97%
N-Ethylbenzylamine
Formula: C₆H₅CH₂NHCH₂CH₃
MW: 135.21 g/mol
Boiling Pt: 168 °C (1013 hPa)
Density: 0.909 g/cm³ (20 °C)
MDL Number: MFCD00009031
CAS Number: 14321-27-8
EINECS: 238-265-1
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR