3-Fluorobenzylamine ≥97%

Danger

Synonyms: (3-Fluorophenyl)methanamine

126896-1G 126896-5G
220035-396EA 25.09 USD
220035-396 220035-397
3-Fluorobenzylamine ≥97%
3-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 87 °C (1013 hPa)
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00008113
CAS Number: 100-82-3
EINECS: 202-891-3
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR