Biphenyl-2,2'-dicarboxylic acid ≥97%

Synonyms: [1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid

126691-25G 126691-100G
220035-381EA 172.62 USD
220035-381 220035-380
Biphenyl-2,2'-dicarboxylic acid ≥97%
Biphenyl-2,2'-dicarboxylic acid
Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
MDL Number: MFCD00002464
CAS Number: 482-05-3
EINECS: 207-576-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR