2,6-Dimethylnaphthalene

126535-1G 126535-5G
220034-921EA 81.95 USD
220034-921 220034-922
2,6-Dimethylnaphthalene
2,6-Dimethylnaphthalene
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 169.8 °C (1013 hPa)
Melting Pt: –44.7 °C
Density: 1.8682 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004120
CAS Number: 581-42-0
EINECS: 209-464-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR