Dimethylammonium chloride

Warning

Synonyms: Dimethylamine hydrochloride

126365-1KG 126365-5G 126365-100G 126365-500G
220035-182EA 122.77 USD
220035-182 220035-184 220035-181 220035-183
Dimethylammonium chloride
Dimethylammonium chloride
Formula: C₂H₈ClN
MW: 81.54 g/mol
Melting Pt: 170…173 °C
MDL Number: MFCD00012477
CAS Number: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR